missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599865
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FCRL4.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein has four extracellular C2-type immunoglobulin domains, a transmembrane domain and a cytoplasmic domain that contains three immune-receptor tyrosine-based inhibitory motifs. This protein may play a role in the function of memory B-cells in the epithelia. Aberrations in the chromosomal region encoding this gene are associated with non-Hodgkin lymphoma and multiple myeloma.
Specifications
| FCRL4 | |
| Polyclonal | |
| Unconjugated | |
| FCRL4 | |
| CD307d; fc receptor homolog 4; Fc receptor like 4; Fc receptor-like 4; Fc receptor-like protein 4; FcRH4; FCRL4; FcR-like protein 4; hIFGP2; IFGP family protein 2; IFGP2; IGFP2; immune receptor translocation-associated protein 1; immunoglobulin superfamily Fc receptor, gp42; immunoglobulin superfamily receptor translocation associated 1; IRTA1; MGC150522; MGC150523 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83417 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96PJ5 | |
| FCRL4 | |
| A synthesized peptide derived from human FCRL4(Accession Q96PJ5), corresponding to amino acid residues T479-S515. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction