missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL5/FCRL3 Antibody (307307), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB20871Y
This item is not returnable.
View return policy
Description
FCRL5/FCRL3 Monoclonal antibody specifically detects FCRL5/FCRL3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FCRL5/FCRL3 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3, Gln16-Arg844, Accession # AAI01067 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 307307 | |
| Flow Cytometry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction