missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL6/FcRH6 Antibody (672505), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6745PECY55
This item is not returnable.
View return policy
Description
FCRL6/FcRH6 Monoclonal antibody specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry
Specifications
| FCRL6/FcRH6 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 343413 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 672505 | |
| Flow Cytometry | |
| Fc receptor-like 6, FLJ16056 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 , Lys16-Leu312, Accession # Q6DN72 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction