missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLA/FCRLM1 Antibody (865910), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7905CL1
This item is not returnable.
View return policy
Description
FCRLA/FCRLM1 Monoclonal antibody specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| FCRLA/FCRLM1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREBFc receptor-like and mucin-like protein 1, MGC4595 | |
| E. coli-derived recombinant mouse FCRLA, Lys221-Lys352, Accession # Q920A9 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 865910 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 84824 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction