missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLA/FCRLM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16462
This item is not returnable.
View return policy
Description
FCRLA/FCRLM1 Polyclonal specifically detects FCRLA/FCRLM1 in Human samples. It is validated for Western Blot.
Specifications
| FCRLA/FCRLM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| FCRLA | |
| Recombinant protein encompassing a sequence within the center region of human FCRLM1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 84824 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREBFc receptor-like and mucin-like protein 1, MGC4595 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction