missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLB/FCRY Antibody (OTI5H6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72384CL1
This item is not returnable.
View return policy
Description
FCRLB/FCRY Monoclonal antibody specifically detects FCRLB/FCRY in Human,Rat samples. It is validated for Western Blot
Specifications
| FCRLB/FCRY | |
| Monoclonal | |
| CoraFluor 1 | |
| Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6 | |
| Human recombinant protein fragment corresponding to amino acids 202-426 of human FCRLB(NP_001002901) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| OTI5H6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 127943 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction