missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLB Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809247
This item is not returnable.
View return policy
Description
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions.Specifications
| FCRLB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6BAA4 | |
| FCRLB | |
| Human recombit protein fragment corresponding to amino acids 202-426 of human FCRLB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| FCRLB | |
| FCRL2, FCRLM2, FCRLY, FREB-2, FREB2, FcRY | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 127943 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction