missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRN/FCGRT Antibody (937508) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8639JF525
This item is not returnable.
View return policy
Description
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| FCRN/FCGRT | |
| Monoclonal | |
| Janelia Fluor 525 | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRN, Ala24-Ser297, Accession # P55899 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 937508 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 2217 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction