missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRN/FCGRT Antibody (CL3640), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 500.00
Specifications
| Antigen | FCRN/FCGRT |
|---|---|
| Clone | CL3640 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209692
|
Novus Biologicals
NBP2-61627 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642238
|
Novus Biologicals
NBP2-61627-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FCRN/FCGRT Monoclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FCRN/FCGRT | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| 2217 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LTFLLFSCPHRLREHLERGRGNLEWKEPPSMRLKARPSSPGFSVLTCSAFSFYPPELQLRFLRNGLAAGTGQGDFGPNSDGSFHASSSLTVKSGDEHHYCCIVQHAGLAQPLRVELESPAKS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL3640 | |
| Monoclonal | |
| Purified | |
| Immunology | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| FCGRT | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title