Learn More
FCRN/FCGRT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-89127
 View more versions of this product
Product Code. 18738733
  • € 550.00 / 0.10mL
Description
FCRN/FCGRT Polyclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
FCGRT
Affinity Purified
RUO
Simple Western reported by internal validation, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
40 kDa
0.1 mL