Learn More
FCRN/FCGRT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-89127
This item is not returnable.
View return policy
Description
FCRN/FCGRT Polyclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| FCGRT | |
| Affinity Purified | |
| RUO |
| Simple Western reported by internal validation, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| 40 kDa | |
| 0.1 mL | |
| IgG |