Learn More
FDFT1 Antibody (OTI2F10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-70715F
Product Code. 18107174
  • € 807.00 / 0.10mL
Description
FDFT1 Monoclonal specifically detects FDFT1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDFT1
Monoclonal
FITC
PBS with 0.05% Sodium Azide
FDFT1
Full length recombinant protein of human FDFT1 (NP_004453) produced in HEK293T cells.
0.1 mL
2222.0
Store at 4°C in the dark.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2F10
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
DGPT, EC 2.5.1.21, ERG9, Farnesyl-diphosphate farnesyltransferase, farnesyl-diphosphate farnesyltransferase 1, FPP:FPP farnesyltransferase, presqualene-di-diphosphate synthase, SQS, squalene synthase, squalene synthetase, SS
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Monkey, Primate
IgG2a