missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDPS Antibody (115), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90812MFV450
This item is not returnable.
View return policy
Description
FDPS Monoclonal antibody specifically detects FDPS in Mouse samples. It is validated for ELISA
Specifications
| FDPS | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 115 | |
| ELISA | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FDPS (Accession#: NP_608219.1; Met1-Lys353). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2224 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction