missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FE65 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-58360MFV610
This item is not returnable.
View return policy
Description
FE65 Polyclonal antibody specifically detects FE65 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| FE65 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein | |
| Recombinant rat FE65, expressed in bacteria. [Swiss-Prot# P46933] | |
| 0.1 mL | |
| Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| 322 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction