Learn More
Eppendorf Feet for CellXpert CS220
Product Code. p-200102295 Shop All Eppendorf Products
Click to view available options
Height (Metric):
2 cm
9 cm
Unit Size:
Each
Product Code. 30213518
Brand: Eppendorf 6740070020
Only null left
Add to basket
Add to basket
Accessory for CellXpert™ CS220 CO2 Incubator Shaker.
  • 1 fixed and 3 height-adjustable
  • For single device or 2 to 3x stack,
  • Choose between 2 cm and 9 cm
  • Eppendorf service required for installation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Feet
Height (English) 0.8 in.
Height (Metric) 2 cm
For Use With (Equipment) CellXpert CS220