missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline CXCR4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4287V-100UG
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Feline samples. It is validated for Flow Cytometry.
Specifications
| CXCR4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P56498 | |
| CXCR4 | |
| HEK293 human embryonic kidney cell line transfected with feline CXCR4 Met1-Ser353 Accession # P56498 | |
| 100 μg | |
| Primary | |
| Detects feline CXCR4. Stains feline CXCR4 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 374606 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7852 | |
| Feline | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction