missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline Fas/TNFRSF6/CD95 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2267S-100UG
This item is not returnable.
View return policy
Description
Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Feline samples. It is validated for Flow Cytometry.
Specifications
| Fas/TNFRSF6/CD95 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001009314 | |
| FAS | |
| Mouse myeloma cell line NS0-derived recombinant feline Fas/TNFRSF6/CD95 Ala25-Lys172 Accession # NP_001009314 | |
| RUO | |
| 355 | |
| Feline | |
| IgG1 |
| Flow Cytometry | |
| 431014 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects feline Fas/TNFRSF6/CD95 in direct ELISAs and Western blots. In direct ELISAs and Western blots no cross-reactivity with recombinant human Fas, recombinant mouse Fas, or recombinant rat Fas are observed | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction