Learn More
R&D Systems™ Feline IFN-gamma ELISpot Development Module, 5 Plate
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
Includes
Feline IFN-gamma Capture Antibody; Feline IFN-gamma Biotinylated-conjugated Detection Antibody
Contains the basic components required to develop an ELISpot (quantitative sandwich ELISA) assay
Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
Generic development protocols provide directi
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |