missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline Immunodeficiency Virus Antibody (NM3-6A9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66500AF350
This item is not returnable.
View return policy
Description
Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Feline Immunodeficiency Virus | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Recognizes Feline Immunodeficieny Virus (FIV) p24 gag derived from p50 GAG polyprotein. This does not recognize Feline Leukemia Virus, Feline Herpes Virus type 1, Feline Coronavirus or Feline Calicivirus. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA | |
| NM3-6A9 | |
| Western Blot, Flow Cytometry, ELISA | |
| CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24 | |
| Native, extracts | |
| Primary | |
| Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction