missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline Immunodeficiency Virus p24 gag Antibody (PAK3-2C1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64802JF646
This item is not returnable.
View return policy
Description
Feline Immunodeficiency Virus p24 gag Monoclonal antibody specifically detects Feline Immunodeficiency Virus p24 gag in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Feline Immunodeficiency Virus p24 gag | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CA, Capsid protein p24, Feline immunodeficiency virus, Feline immunodeficiency virus capsid protein p24, Feline immunodeficiency virus p24, FIV capsid protein p24 | |
| Made to Viral FELINE IMMUNODEFICIENCY VIRUS p24 gag | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| PAK3-2C1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction