Learn More
Feline leukemia virus p27 Mouse anti-Virus, Clone: HM189, Abnova™
Mouse Monoclonal Antibody
Brand:  Abnova MAB13548.500ug
Product Code. 16092066
  • € 877.00 / 500µg
Description
Mouse monoclonal antibody raised against feline leukemia virus p27.
  • High specificity
  • High reactivity
Specifications
Specifications
Feline leukemia virus p27
Monoclonal
Unconjugated
ELISA Western Blot The optimal working dilution should be determined by the end user.
Mouse
500 μg
Primary
Virus
Liquid
ELISA, Western Blot
HM189
Mouse monoclonal antibody raised against feline leukemia virus p27.
In PBS, pH 7.2 (0.09% sodium azide).
Protein A purification
RUO
This antibody reacts with p27 of feline leukemia virus.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only (RUO)