Learn More
Value Plastics Female Luer to 300 Series Barb
Product Code. 30054362 Shop All Cole-Parmer Products
Click to view available options
Hose Barb:
1/4 in.
Unit Size:
Pack of 25
Product Code. 30054362
Brand: Value Plastics 5012069
Only null left
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Material Polypropylene
Packaging Quantity 25/Pk.
Hose Barb 1/4 in.
Inlet Female Luer, Hose Barb
Always test the application to verify the performance of the interconnection.