Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Elbow
Shop All Cole-Parmer Products
Click to view available options
Material:
Nylon
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Constructed of durable chemical resistance nylon. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Inlet | Female Luer Thread, 1/8-27 NPT(M) Thread |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.