Learn More
Value Plastics Female Luer Thread Union Fittings
Product Code. p-200105066 Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Product Code. 30053660
Brand: Value Plastics 5012066
Only null left
Add to basket
Add to basket
Luer fittings allow for a simple yet secure connection
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;FDA
Packaging Quantity 25/Pk.
Inlet Female Luer Thread
Material (Seal) Polycarbonate