missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Antibody (OTI1F3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70716CL1
This item is not returnable.
View return policy
Description
FEN-1 Monoclonal antibody specifically detects FEN-1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| FEN-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| Human recombinant protein fragment corresponding to amino acids 117-380 of human FEN1 (NP_004102) produced in E.coli. | |
| 0.1 mL | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| 2237 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1F3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction