missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Antibody (S01-3I5), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | FEN-1 |
|---|---|
| Clone | S01-3I5 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625083
|
Novus Biologicals
NBP3-19863-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687123
|
Novus Biologicals
NBP3-19863-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FEN-1 Monoclonal antibody specifically detects FEN-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FEN-1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| A synthetic peptide of human FEN-1 (Uniprot # P39748) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-3I5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 2237 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title