missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FEN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582261
This item is not returnable.
View return policy
Description
Immunogen sequence: CAALVKAGKV YAAATEDMDC LTFGSPVLMR HLTASEAKKL PIQEFHLSRI LQELGLNQEQ FVDLCILLGS DYCESIRGIG PKRAVDLIQK HKSIEEIVRR LDPNKYPVPE NWLHKEAHQL FLEPEVLDPE SVELKW Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 97%.
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
Specifications
| FEN1 | |
| Polyclonal | |
| Unconjugated | |
| FEN1 | |
| AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14156, 2237, 84490 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P39748, P39749, Q5XIP6 | |
| FEN1 | |
| Recombinant protein corresponding to Human FEN1. Recombinant protein control fragment (Product #RP-89493). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction