Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Fenazaquin |
| CAS | 120928-09-8 |
| CAS Max % | ≤ 100% |
| Density | 1.16 g/cm3 |
| Beilstein | 8331263 |
| Color | Colorless |
| Melting Point | 80.5°C |
| Boiling Point | 307°C (Decomposition) |
| Synonym | 4-tert-butylphenethyl quinazolin-4-yl ether |
| Molecular Weight (g/mol) | 306.40 g/mol |
| Show More |