Learn More
Fenazaquin, TRC
Specifications
Specifications
| Chemical Name or Material | Fenazaquin |
| CAS | 120928-09-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H22 N2 O |
| Quantity | 10 g |
| Synonym | 4-[2-[4-(1,1-Dimethylethyl)phenyl]ethoxy]quinazoline, EL 436, Fenaza, Fenazachin, Fenazaquin, GWN 1708, Magister, Magus, Phenazaquin, Pride Ultra, XDE 436 |
| InChI Formula | InChI=1S/C20H22N2O/c1-20(2,3)16-10-8-15(9-11-16)12-13-23-19-17-6-4-5-7-18(17)21-14-22-19/h4-11,14H,12-13H2,1-3H3 |
| SMILES | CC(C)(C)c1ccc(CCOc2ncnc3ccccc23)cc1 |
| IUPAC Name | 4-[2-(4-tert-butylphenyl)ethoxy]quinazoline |
| Molecular Weight (g/mol) | 306.4 |
| Show More |