Learn More
Fenebrutinib, MedChemExpress
Product Code. 16490398
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16490398
Brand: Medchem Express HY19834
MedChemExpress Fenebrutinib (GDC-0853) is a potent, selective, orally available, and noncovalent bruton's tyrosine kinase (Btk) inhibitor with Kis of 0.91 nM, 1.6, 1.3, 12.6, and 3.4 nM for WT Btk, and the C481S, C481R, T474I, T474M mutants. Fenebrutinib has the potential for rheumatoid arthritis and systemic lupus erythematosus research.
Fenebrutinib (GDC-0853) is a potent, selective, orally available, and noncovalent bruton's tyrosine kinase (Btk) inhibitor with Kis of 0.91 nM, 1.6, 1.3, 12.6, and 3.4 nM for WT Btk, and the C481S, C481R, T474I, T474M mutants. Fenebrutinib has the potential for rheumatoid arthritis and systemic lupus erythematosus research.
Specifications
Chemical Name or Material Fenebrutinib
CAS 1434048-34-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C37H44N8O4
Quantity 5 mg
Synonym GDC-0853
Solubility Information DMSO : ≥ 23 mg/mL (34.60 mM)
SMILES O=C1C(NC(C=C2)=NC=C2N3CCN(C4COC4)C[C@@H]3C)=CC(C5=CC=NC(N6C(C7=CC(CC(C)(C)C8)=C8N7CC6)=O)=C5CO)=CN1C
Molecular Weight (g/mol) 664.8
Formula Weight 664.8
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only