missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FENS1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-921
This item is not returnable.
View return policy
Description
FENS1 Polyclonal specifically detects FENS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| FENS1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FENS-1Zinc finger FYVE domain-containing protein 17, KIAA1435, Phosphoinositide-binding protein 1, WD repeat and FYVE domain containing 1, WD repeat and FYVE domain-containing protein 1, WD40 and FYVE domain containing 1, WDF1WD40- and FYVE domain-containing protein 1, ZFYVE17phosphoinositide-binding protein SR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57590 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 4 - 6 ug/ml, Immunohistochemistry-Paraffin 4 - 6 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_065881 | |
| WDFY1 | |
| Peptide with sequence VVGCSLATGFSPH corresponding to C-Terminus according to NP_065881.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (as represented by NP_065881 and NP_848127) | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction