Learn More
Versilon FEP High-Purity Tubing
Product Code. 30053362 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
Diameter (English) Outer:
1 in.
1-1/8 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
Unit Size:
15m
Product Code. 30053362
Brand: Versilon 5010725
Only null left
Add to basket
Add to basket
High-performance tubing exhibits excellent physical and chemical resistance
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Made from chemically inert thermoplastic. Transparent and flexible—ideal for chemical transfer applications. Withstands temperatures up to 206°C (402°F). Noncorroding, nonoxidizing, and UV resistant
Specifications
Certifications/Compliance ADCF
Diameter (English) Inner 1/8 in.
Material FEP
Diameter (English) Outer 1/4 in.
Overall Length (Metric) 50 ft.
Color Transparent
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.