Learn More
Saint-Gobain FEP Tubing
Product Code. 11757335
Click to view available options
Diameter (Metric) Inner:
0.8 mm
Unit Size:
10m
Product Code. 11757335
Brand: Saint-Gobain C20080X04010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance FDA compliant (food), USP Class VI, manufactured according to GLP
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm
Material FEP
Diameter (Metric) Outer 1.6 mm
Overall Length (Metric) 10 m
Thickness (Metric) 0.4 mm
For Use With (Application) Transfer fluid pressure or high temperature in the chemical, pharmaceutical, mechanical, electrical, automobile
Min. Operating Temperature (Metric) -270°C
Packaging Quantity 1 Each
Sterility Sterile
Color Translucent
Flexibility Semirigid
Maximum Pressure 34 bar
Max. Operating Temperature (Metric) 205°C