missing translation for 'onlineSavingsMsg'
Learn More

Feprazone, Mikromol™

Product Code. 17990416
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 17990416

Brand: Mikromol MM0691.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Feprazone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Feprazone
CAS 30748-29-9
Analyte or Component Names Feprazone
Shipping Condition Room Temperature
Molecular Formula C20 H20 N2 O2
Certifications/Compliance ISO 17025
Synonym 3,5-Pyrazolidinedione, 4-(3-methyl-2-buten-1-yl)-1,2-diphenyl-, 3,5-Pyrazolidinedione, 4-(3-methyl-2-butenyl)-1,2-diphenyl- (8CI,9CI), 4-(2-Isopentenyl)-1,2-diphenyl-3,5-pyrazolidinedione, 4-(3-Methyl-2-butenyl)-1,2-diphenyl-3,5-pyrazolidinedione, 4-Prenyl-1,2-diphenyl-3,5-dioxopyrazolidine, 4-Prenyl-1,2-diphenyl-3,5-pyrazolidinedione, Analud, DA 2370, Fenilprenazone, Feprazone, Methrazone, Metrazone, Phenylprenazone, Pheprazone, Prenazone, Zepelin
InChI Formula InChI=1S/C20H20N2O2/c1-15(2)13-14-18-19(23)21(16-9-5-3-6-10-16)22(20(18)24)17-11-7-4-8-12-17/h3-13,18H,14H2,1-2H3
SMILES CC(=CCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3)C
IUPAC Name 4-(3-methylbut-2-enyl)-1,2-diphenylpyrazolidine-3,5-dione
Molecular Weight (g/mol) 320.38
Quantity 50 mg
Formula Weight 320.1525 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.