Learn More
FER Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-25287-100UL
Description
The FER antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FER. This antibody recognizes human, mouse antigen. The FER antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
| FER | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 2241 | |
| IgG |