missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FER1L5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05642-20ul
This item is not returnable.
View return policy
Description
FER1L5 Polyclonal antibody specifically detects FER1L5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| FER1L5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| fer-1-like 5 (C. elegans), fer-1-like protein 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1350-1670 of human FER1L5 (NP_001280012.1). KLPTLSEKKHQDFLGYLYRKFWFKSSKAEDEYEHEVDWWSKLFWATDEHKSLKYKYKDYHTLKVYECELEAVPAFQGLQDFCQTFKLYQEQPKLDSPVVGEFKGLFRIYPFPENPEAPKPPLQFLVWPEREDFPQPCLVRVYMVRAINLQPQDYNGLCDPYVILKLGKTELGNRDMYQPNTLDPIFGMMFELTCNIPLEKDLEIQLYDFDLFSPDDKIGTTVIDLENRLLSGFGAHCGLSKSYCQSGPFRWRDQMPPSYLLERYAKRKGLPPPLFSPEEDAVFYNGKKFKLQSFEPKTPTVHGLGPKKERLALYLLHTQGL | |
| 20 μg | |
| Cell Biology | |
| 90342 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction