missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FERD3L Polyclonal Antibody
GREENER_CHOICE

Product Code. 15758221 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15758221 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15758221 Supplier Invitrogen™ Supplier No. PA560389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAAYPESCVD TTVLDFVADL SLASPRRPLL CDFAPGVSLG DPALALREGR PRRMARFEEG DPEEEECEVD Q Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 56%.

FERD3L is an evolutionarily conserved bHLH transcription factor that is identified in fly, mouse and man. Nervous system expression of this protein is detected in fly and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FERD3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FERD3L
Gene Accession No. Q96RJ6
Gene Alias Basic helix-loop-helix protein N-twist; bHLHa31; Class A basic helix-loop-helix protein 31; fer3; Fer3-like (Drosophila); Fer3-like bHLH transcription factor; fer3-like protein; FERD3L; Mnato3; Nato; NATO3; nephew of atonal 3; Neuronal twist; NTWIST; N-TWIST
Gene Symbols FERD3L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FERD3L. Recombinant protein control fragment (Product #RP-98483).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.