Learn More
Thermo Scientific™ FaqI (BsmFI) (2 U/μL)
Cut at GGGAC(10/14)^ sites with FaqI (BsmFI) restriction enzyme, which performs best at 37°C in Tango (+SAM) buffer (Isoschizomers: BslFI, BsmFI).
Brand:  Thermo Scientific™ ER1811
64.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Product Code. 10850041
  • € 96.00 / 100 units
In Stock
to see stock.
Description
Description
Lambda DNA digested with FaqI (BsmFI), 1% agarose, 38 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications
Specifications
2 U/μL
FaqI (BsmFI)
Yes
Yes
Scarless Cloning
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer Tango
37°C
Restriction Enzyme