Learn More
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker
Perform sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels.
Brand: Thermo Scientific™ SM0221
Additional Details : Weight : 1.51950kg
Description
pUC19 DNA/MspI (HpaII) Marker is recommended for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
pUC19 DNA is digested to completion with the appropriate Fermentas PureExtreme restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the marker's preparation. pUC19 DNA/MspI (HpaII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
6X | |
DNA/MSPI (HPAII) Marker | |
Agarose Gels | |
pUC19 DNA/MspI (HpaII) Marker |
Agarose | |
No | |
Sustainable packaging | |
50 μg |
For Research Use Only. Not for use in diagnostic procedures.