missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FERMT3/URP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76349AF350
This item is not returnable.
View return policy
Description
FERMT3/URP2 Polyclonal antibody specifically detects FERMT3/URP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| FERMT3/URP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 350 | |
| fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of the human KINDLIN3. The immunogen is located within the last 50 amino acids of KINDLIN3. Amino Acid Squence: STRERARGEELDEDLFL | |
| 0.1 mL | |
| Signal Transduction | |
| 83706 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction