missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Antibody (39G3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 533.00
Specifications
| Antigen | Ferritin |
|---|---|
| Clone | 39G3 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043541
|
Novus Biologicals
NBP3-26567-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043812
|
Novus Biologicals
NBP3-26567-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Ferritin | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| A synthesized peptide derived from Human Ferritin [UniProt P02794] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 39G3 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2495 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title