missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Heavy Chain Antibody (962610), Alexa Fluor™ 532, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB9354AF532
This item is not returnable.
View return policy
Description
Ferritin Heavy Chain Monoclonal specifically detects Ferritin Heavy Chain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Ferritin Heavy Chain | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FTH1 | |
| The immunogen is raised against Ferritin Heavy Chain. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 962610 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Mouse | |
| 0.1 mL | |
| 2495 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction