missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Heavy Chain Antibody (962610), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB9354AF700
This item is not returnable.
View return policy
Description
Ferritin Heavy Chain Monoclonal antibody specifically detects Ferritin Heavy Chain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry
Specifications
| Ferritin Heavy Chain | |
| Monoclonal | |
| Alexa Fluor 700 | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| The immunogen is raised against Ferritin Heavy Chain. | |
| 100 μg | |
| Lipid and Metabolism | |
| 2495 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
| 962610 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction