missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Heavy Chain Antibody (FTH/8700R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20850APC
This item is not returnable.
View return policy
Description
Ferritin Heavy Chain Monoclonal antibody specifically detects Ferritin Heavy Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ferritin Heavy Chain | |
| Monoclonal | |
| APC | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant human Ferritin Heavy Chain protein fragment (around aa 58-180) (exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2495 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FTH/8700R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction