missing translation for 'onlineSavingsMsg'
Learn More

Ferritin Heavy Chain Antibody (FTH/8700R), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20850C

Product Code. 18579287

  • € 762.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ferritin Heavy Chain Monoclonal antibody specifically detects Ferritin Heavy Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ferritin Heavy Chain
Monoclonal
DyLight 650
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Recombinant human Ferritin Heavy Chain protein fragment (around aa 58-180) (exact sequence is proprietary)
0.1 mL
Lipid and Metabolism
2495
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
FTH/8700R
50 mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.