missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin, Light Chain Mouse, Clone: FTL/1388, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | Ferritin, Light Chain |
|---|---|
| Clone | FTL/1388 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15980058
|
EnquireBio
2512-MSM3-P0 |
20 μg | |||||||
|
15990058
|
EnquireBio
2512-MSM3-P1 |
100 μg |
€ 573.00 € 478.00 / 100µg Save € 95.00 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Ferritin, Light Chain | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 2512 | |
| Recombinant human FTL protein fragment (aa 38-165) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 19-25kDa |
| FTL/1388 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_000137 | |
| FTL | |
| IgG2a | |
| Protein A/G Purified | |
| Cytoplasmic | |
| HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title