Learn More
Ferroportin/SLC40A1 Antibody (1308C), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB9924J
Product Code. 18076525
  • € 663.00 / 0.10mL
Description
Ferroportin/SLC40A1 Monoclonal antibody specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferroportin/SLC40A1
Monoclonal
Janelia Fluor 646
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1
Synthetic peptide containing human Ferroportin/SLC40A1, Accession # Q9NP59
0.1 mL
Cancer, Lipid and Metabolism, Signal Transduction
30061
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunohistochemistry
1308C
50mM Sodium Borate
Rabbit
Protein A or G purified from hybridoma culture supernatant