Learn More
Novus Biologicals™ Ferroportin/SLC40A1 Antibody (Amgen patent anti-Ferroportin) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28538-100ug
 View more versions of this product
Product Code. 30047480
  • € 338.00 / 100µg
Description
Ferroportin/SLC40A1 Monoclonal antibody specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferroportin/SLC40A1
Monoclonal
Unconjugated
Q9NP59
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG
Flow Cytometry, ELISA, Functional Assay
Amgen patent anti-Ferroportin
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1
SLC40A1
100 μg
Cancer, Lipid and Metabolism, Signal Transduction
30061