missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ FerroVer™ Iron Reagent Powder Pillows

Product Code. 10395822
Change view
Click to view available options
Quantity:
10 mL
100 Pack
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10395822 10 mL Pack of 100
10530213 100 Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10395822 Supplier Hach Lange™ Supplier No. 2105769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hach Lange FerroVer Iron Reagent Powder Pillows, Quantity: 10mL, Physical Form: Solid

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 10 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • FerroVer Iron Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Sodium thiosulfate,sodium metabisulphite, sodium dithionite, sodium hydrosulphite, 1, 10-Phenanthro
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.