missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FES Recombinant Rabbit Monoclonal Antibody (K01_1R12)
GREENER_CHOICE

Product Code. p-200114235
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829175 100 μL
30827478 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829175 Supplier Invitrogen™ Supplier No. MA563515

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

FES Recombinant Monoclonal Antibody for Western Blot

FES (FPS) is a tyrosine protein kinase that is required for maintenance of cellular transformation. Its chromosomal location is linked to a specific translocation event identified in patients with acute promyelocytic leukemia, and it is also involved in normal hematopoiesis. Alternative splicing results in multiple variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FES
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1R12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene FES
Gene Accession No. P07332
Gene Alias AI586313; BB137047; c-fes; EC 2.7.10.2; feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; feline sarcoma/Fujinami avian sarcoma oncogene homolog; Fes; FES proto-oncogene, tyrosine kinase; FPS; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; RGD1564385; tyrosine kinase Fps/Fes; tyrosine-protein kinase Fes/Fps; Variant 4
Gene Symbols FES
Host Species Rabbit
Immunogen A synthetic peptide of human FES.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2242
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.