missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetal Hemoglobin Antibody, Alexa Fluor™ 532 , Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB110-41084AF532
This item is not returnable.
View return policy
Description
Fetal Hemoglobin Polyclonal antibody specifically detects Fetal Hemoglobin in Human samples. It is validated for Western Blot, ELISA
Specifications
| Fetal Hemoglobin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| 3047 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 532 | |
| abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979 | |
| Purified human hemoglobin F of fetal cord blood cell origin. | |
| 1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction